ADVH49B832C8 AChemBlock

6-bromo-7-methylchroman-4-one

  • Cas Number : 173381-62-9
  • Molecular Weight : 241.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-7-methylchroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87677
Availability details for
Quantity
  • Availability
  • Add to Cart