ADVH49B832CA AChemBlock

7-chloro-3,4-dihydronaphthalen-2(1H)-one

  • Cas Number : 17556-19-3
  • Molecular Weight : 180.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-3-4-dihydronaphthalen-2-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87679
Availability details for
Quantity
  • Availability
  • Add to Cart