ADVH49B832CE AChemBlock

3-bromo-5-(piperidin-2-yl)pyridine

  • Cas Number : 179119-97-2
  • Molecular Weight : 241.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-piperidin-2-yl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87683
Availability details for
Quantity
  • Availability
  • Add to Cart