ADVH49B832D0 AChemBlock

(R)-1-(4-((trifluoromethyl)thio)phenyl)ethan-1-amine hydrochloride

  • Cas Number : 1800240-39-4
  • Molecular Weight : 257.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-4-trifluoromethyl-thio-phenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87685
Availability details for
Quantity
  • Availability
  • Add to Cart