ADVH49B832D1 AChemBlock

tert-butyl (1-(3-aminophenyl)ethyl)carbamate

  • Cas Number : 180079-59-8
  • Molecular Weight : 236.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-3-aminophenyl-ethyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87686
Availability details for
Quantity
  • Availability
  • Add to Cart