ADVH49B832D2 AChemBlock

(6-bromo-2-methoxypyridin-3-yl)methanamine

  • Cas Number : 1802489-58-2
  • Molecular Weight : 217.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-methoxypyridin-3-yl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87687
Availability details for
Quantity
  • Availability
  • Add to Cart