ADVH49B832D3 AChemBlock

(6-bromo-2-methoxypyridin-3-yl)methanol

  • Cas Number : 1802489-60-6
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-methoxypyridin-3-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87688
Availability details for
Quantity
  • Availability
  • Add to Cart