ADVH49B832D4 AChemBlock

5-bromo-2-(pyrrolidin-2-yl)pyridine dihydrochloride

  • Cas Number : 1803590-14-8
  • Molecular Weight : 300.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-pyrrolidin-2-yl-pyridine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87689
Availability details for
Quantity
  • Availability
  • Add to Cart