ADVH49B832DC AChemBlock

5,8-difluorochroman-4-amine hydrochloride

  • Cas Number : 1810070-13-3
  • Molecular Weight : 221.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-8-difluorochroman-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87697
Availability details for
Quantity
  • Availability
  • Add to Cart