ADVH49B832DD AChemBlock

1-(5-methylfuran-2-yl)propan-1-amine hydrochloride

  • Cas Number : 1810070-14-4
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-5-methylfuran-2-yl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87698
Availability details for
Quantity
  • Availability
  • Add to Cart