ADVH49B832DF AChemBlock

(R)-6-(trifluoromethoxy)chroman-4-amine hydrochloride

  • Cas Number : 1810074-61-3
  • Molecular Weight : 269.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-6-trifluoromethoxy-chroman-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87700
Availability details for
Quantity
  • Availability
  • Add to Cart