ADVH49B832E8 AChemBlock

(R)-2-amino-2-(3,4-dichlorophenyl)ethan-1-ol hydrochloride

  • Cas Number : 1810074-84-0
  • Molecular Weight : 242.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-2-3-4-dichlorophenyl-ethan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87709
Availability details for
Quantity
  • Availability
  • Add to Cart