ADVH49B832EF AChemBlock

7-methylchroman-4-one

  • Cas Number : 18385-69-8
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-methylchroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87716
Availability details for
Quantity
  • Availability
  • Add to Cart