ADVH49B832F0 AChemBlock

7-chlorochroman-4-one

  • Cas Number : 18385-72-3
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-chlorochroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87717
Availability details for
Quantity
  • Availability
  • Add to Cart