ADVH49B832F1 AChemBlock

(S)-3-((tert-butoxycarbonyl)amino)pentanoic acid

  • Cas Number : 183990-48-9
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-tert-butoxycarbonyl-amino-pentanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87718
Availability details for
Quantity
  • Availability
  • Add to Cart