ADVH49B832F3 AChemBlock

2-methoxy-5-(pyrrolidin-2-yl)pyridine

  • Cas Number : 185510-44-5
  • Molecular Weight : 178.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-5-pyrrolidin-2-yl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87720
Availability details for
Quantity
  • Availability
  • Add to Cart