ADVH49B832F5 AChemBlock

1-(4-chlorophenyl)-2-methylpropan-1-one

  • Cas Number : 18713-58-1
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-chlorophenyl-2-methylpropan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87722
Availability details for
Quantity
  • Availability
  • Add to Cart