ADVH49B832F7 AChemBlock

2-amino-2-(3,4-dichlorophenyl)ethan-1-ol

  • Cas Number : 188586-38-1
  • Molecular Weight : 206.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-2-3-4-dichlorophenyl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87724
Availability details for
Quantity
  • Availability
  • Add to Cart