ADVH49B832F8 AChemBlock

2-(pyrrolidin-2-yl)phenol hydrochloride

  • Cas Number : 1894060-68-4
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-pyrrolidin-2-yl-phenol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87725
Availability details for
Quantity
  • Availability
  • Add to Cart