ADVH49B832FD AChemBlock

8-fluorochroman-4-amine hydrochloride

  • Cas Number : 191608-18-1
  • Molecular Weight : 203.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-fluorochroman-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87730
Availability details for
Quantity
  • Availability
  • Add to Cart