ADVH49B832FF AChemBlock

6-fluorochroman-4-amine hydrochloride

  • Cas Number : 191609-45-7
  • Molecular Weight : 203.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-fluorochroman-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87732
Availability details for
Quantity
  • Availability
  • Add to Cart