ADVH49B83305 AChemBlock

2-(aminomethyl)-6-methylphenol hydrochloride

  • Cas Number : 1956330-87-2
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-aminomethyl-6-methylphenol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87738
Availability details for
Quantity
  • Availability
  • Add to Cart