ADVH49B83306 AChemBlock

1-(3-chloro-2-fluorophenyl)ethan-1-amine hydrochloride

  • Cas Number : 1956371-30-4
  • Molecular Weight : 210.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-chloro-2-fluorophenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87739
Availability details for
Quantity
  • Availability
  • Add to Cart