ADVH49B83307 AChemBlock

3-(2,3-dichlorophenyl)pyrrolidine hydrochloride

  • Cas Number : 1956386-37-0
  • Molecular Weight : 252.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-3-dichlorophenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87740
Availability details for
Quantity
  • Availability
  • Add to Cart