ADVH49B83308 AChemBlock

(S)-2-amino-3-(4-chlorophenyl)propan-1-ol hydrochloride

  • Cas Number : 1956434-75-5
  • Molecular Weight : 222.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-3-4-chlorophenyl-propan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87741
Availability details for
Quantity
  • Availability
  • Add to Cart