ADVH49B83309 AChemBlock

(S)-3-amino-3-cyclopentylpropanoic acid hydrochloride

  • Cas Number : 1956435-02-1
  • Molecular Weight : 193.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-amino-3-cyclopentylpropanoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87742
Availability details for
Quantity
  • Availability
  • Add to Cart