ADVH49B8330B AChemBlock

(S)-7-methylchroman-4-amine hydrochloride

  • Cas Number : 1956436-49-9
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-7-methylchroman-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87744
Availability details for
Quantity
  • Availability
  • Add to Cart