ADVH49B8330C AChemBlock

(S)-2-(2,2-difluorobenzo[d][1,3]dioxol-4-yl)pyrrolidine hydrochloride

  • Cas Number : 1956437-18-5
  • Molecular Weight : 263.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-2-2-difluorobenzo-d-1-3-dioxol-4-yl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87745
Availability details for
Quantity
  • Availability
  • Add to Cart