ADVH49B8331C AChemBlock

(R)-2-amino-2-(naphthalen-2-yl)ethan-1-ol

  • Cas Number : 204851-80-9
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-2-naphthalen-2-yl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87761
Availability details for
Quantity
  • Availability
  • Add to Cart