ADVH49B8331F AChemBlock

(R)-3-(1-amino-2,2,2-trifluoroethyl)benzoic acid hydrochloride

  • Cas Number : 2055848-93-4
  • Molecular Weight : 255.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-1-amino-2-2-2-trifluoroethyl-benzoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87764
Availability details for
Quantity
  • Availability
  • Add to Cart