ADVH49B83327 AChemBlock

2-amino-2-(3,5-dichlorophenyl)ethan-1-ol hydrochloride

  • Cas Number : 2061980-48-9
  • Molecular Weight : 242.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-2-3-5-dichlorophenyl-ethan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87772
Availability details for
Quantity
  • Availability
  • Add to Cart