ADVH49B8332B AChemBlock

(R)-4-(1-aminoethyl)phenol hydrochloride

  • Cas Number : 2061996-43-6
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-4-1-aminoethyl-phenol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87776
Availability details for
Quantity
  • Availability
  • Add to Cart