ADVH49B83330 AChemBlock

(R)-1-(6-fluoropyridin-2-yl)ethan-1-amine dihydrochloride

  • Cas Number : 2061996-65-2
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-6-fluoropyridin-2-yl-ethan-1-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87781
Availability details for
Quantity
  • Availability
  • Add to Cart