ADVH49B83332 AChemBlock

(R)-4-(1-amino-2-methoxyethyl)benzonitrile hydrochloride

  • Cas Number : 2061996-79-8
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-4-1-amino-2-methoxyethyl-benzonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87783
Availability details for
Quantity
  • Availability
  • Add to Cart