ADVH49B83333 AChemBlock

tert-butyl (S)-2-(3-bromophenyl)pyrrolidine-1-carboxylate

  • Cas Number : 2061996-90-3
  • Molecular Weight : 326.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-s-2-3-bromophenyl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87784
Availability details for
Quantity
  • Availability
  • Add to Cart