ADVH49B83336 AChemBlock

1-(2-fluoro-5-methoxyphenyl)ethan-1-amine hydrochloride

  • Cas Number : 2089380-99-2
  • Molecular Weight : 205.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-fluoro-5-methoxyphenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87787
Availability details for
Quantity
  • Availability
  • Add to Cart