ADVH49B8333A AChemBlock

(S)-2-amino-2-(3-bromophenyl)ethan-1-ol hydrochloride

  • Cas Number : 2095772-98-6
  • Molecular Weight : 252.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-2-3-bromophenyl-ethan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87791
Availability details for
Quantity
  • Availability
  • Add to Cart