ADVH49B8333D AChemBlock

1-(pyridin-4-yl)butan-1-amine dihydrochloride

  • Cas Number : 2097938-67-3
  • Molecular Weight : 223.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-pyridin-4-yl-butan-1-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87794
Availability details for
Quantity
  • Availability
  • Add to Cart