ADVH49B8333E AChemBlock

methyl (R)-3-amino-3-(3,4-dichlorophenyl)propanoate hydrochloride

  • Cas Number : 2097958-02-4
  • Molecular Weight : 284.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-r-3-amino-3-3-4-dichlorophenyl-propanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87795
Availability details for
Quantity
  • Availability
  • Add to Cart