ADVH49B83340 AChemBlock

(S)-4-methyl-2,3-dihydro-1H-inden-1-amine hydrochloride

  • Cas Number : 2109874-04-4
  • Molecular Weight : 183.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-4-methyl-2-3-dihydro-1h-inden-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87797
Availability details for
Quantity
  • Availability
  • Add to Cart