ADVH49B83342 AChemBlock

4-((carboxymethyl)amino)benzoic acid hydrochloride

  • Cas Number : 2109874-13-5
  • Molecular Weight : 231.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-carboxymethyl-amino-benzoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87799
Availability details for
Quantity
  • Availability
  • Add to Cart