ADVH49B83343 AChemBlock

2,3-dihydrothieno[3,4-b][1,4]dioxine-5,7-dicarbaldehyde

  • Cas Number : 211235-87-9
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-dihydrothieno-3-4-b-1-4-dioxine-5-7-dicarbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87800
Availability details for
Quantity
  • Availability
  • Add to Cart