ADVH49B83344 AChemBlock

(R)-2-amino-2-(5-bromopyridin-2-yl)ethan-1-ol dihydrochloride

  • Cas Number : 2126088-18-2
  • Molecular Weight : 289.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-2-5-bromopyridin-2-yl-ethan-1-ol-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87801
Availability details for
Quantity
  • Availability
  • Add to Cart