ADVH49B83346 AChemBlock

3-amino-3-(2,3-dimethoxyphenyl)propan-1-ol hydrochloride

  • Cas Number : 2135331-86-9
  • Molecular Weight : 247.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-3-2-3-dimethoxyphenyl-propan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87803
Availability details for
Quantity
  • Availability
  • Add to Cart