ADVH49B83348 AChemBlock

2,2,2-trifluoro-1-(pyridin-4-yl)ethan-1-amine dihydrochloride

  • Cas Number : 2153472-94-5
  • Molecular Weight : 249.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-2-trifluoro-1-pyridin-4-yl-ethan-1-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87805
Availability details for
Quantity
  • Availability
  • Add to Cart