ADVH49B8334B AChemBlock

(S)-2-(3,4-difluorophenyl)pyrrolidine hydrochloride

  • Cas Number : 2177258-16-9
  • Molecular Weight : 219.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-3-4-difluorophenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87808
Availability details for
Quantity
  • Availability
  • Add to Cart