ADVH49B83353 AChemBlock

(S)-2,2,2-trifluoro-1-(naphthalen-1-yl)ethan-1-amine hydrochloride

  • Cas Number : 2222662-58-8
  • Molecular Weight : 261.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-2-2-trifluoro-1-naphthalen-1-yl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87816
Availability details for
Quantity
  • Availability
  • Add to Cart