ADVH49B83354 AChemBlock

(R)-6-bromo-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • Cas Number : 2241594-26-1
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-6-bromo-1-2-3-4-tetrahydronaphthalen-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87817
Availability details for
Quantity
  • Availability
  • Add to Cart