ADVH49B83357 AChemBlock

(R)-3-(pyrrolidin-2-yl)phenol hydrochloride

  • Cas Number : 2241594-46-5
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-pyrrolidin-2-yl-phenol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87820
Availability details for
Quantity
  • Availability
  • Add to Cart