ADVH49B8335B AChemBlock

(R)-5-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • Cas Number : 2250243-14-0
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-5-methyl-1-2-3-4-tetrahydronaphthalen-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87824
Availability details for
Quantity
  • Availability
  • Add to Cart